Product Details

SNP ID
rs9992501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174927432 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAACTGGCACCTTTACTATGTTGA[A/C]TGTTCTAATCTATCTGTATTTGATT
Phenotype
MIM: 604778
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADAM29 PubMed Links

Gene Details

Gene
ADAM29
Gene Name
ADAM metallopeptidase domain 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130703.1 Intron NP_001124175.1
NM_001130704.1 Intron NP_001124176.1
NM_001130705.1 Intron NP_001124177.1
NM_001278125.1 Intron NP_001265054.1
NM_001278126.1 Intron NP_001265055.1
NM_001278127.1 Intron NP_001265056.1
NM_014269.4 Intron NP_055084.3
XM_011531556.1 Intron XP_011529858.1
XM_011531557.1 Intron XP_011529859.1
XM_011531559.1 Intron XP_011529861.1
XM_011531560.1 Intron XP_011529862.1
XM_011531561.1 Intron XP_011529863.1

View Full Product Details