Product Details

SNP ID
rs7755203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118503165 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTCAACAATGAACAAGTGGAATA[C/T]AAAAATTAAAAAATTCACTACCATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRD7P3 PubMed Links

Gene Details

Gene
BRD7P3
Gene Name
bromodomain containing 7 pseudogene 3
There are no transcripts associated with this gene.

Gene
CEP85L
Gene Name
centrosomal protein 85 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042475.2 Intron NP_001035940.1
NM_001178035.1 Intron NP_001171506.1
NM_206921.2 Intron NP_996804.2
XM_005266970.1 Intron XP_005267027.1
XM_005266971.1 Intron XP_005267028.1
XM_005266972.4 Intron XP_005267029.1
XM_006715475.3 Intron XP_006715538.1
XM_011535809.2 Intron XP_011534111.1
XM_011535810.2 Intron XP_011534112.1
XM_011535811.1 Intron XP_011534113.1
XM_017010846.1 Intron XP_016866335.1
XM_017010847.1 Intron XP_016866336.1

View Full Product Details