Product Details
- SNP ID
-
rs7146028
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:52736721 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGAATTTTTTAAAAATACTGATGC[C/G]TGGACCCTACCAGCACCTATTGTAG
- Phenotype
-
MIM: 602708
MIM: 615814
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PSMC6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78402324] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PSMC6
- Gene Name
- proteasome 26S subunit, ATPase 6
There are no transcripts associated with this gene.
- Gene
- STYX
- Gene Name
- serine/threonine/tyrosine interacting protein
View Full Product Details