Product Details

SNP ID
hCV44804184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105491662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCCTCTTCCGTGTGCTCTCGC[C/T]ACTCCCTGCGGGCAACGCCTTGGCA
Phenotype
MIM: 123875
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf80 PubMed Links

Gene Details

Gene
C14orf80
Gene Name
chromosome 14 open reading frame 80
There are no transcripts associated with this gene.

Gene
CRIP1
Gene Name
cysteine rich protein 1
There are no transcripts associated with this gene.

View Full Product Details