Product Details

SNP ID
rs7974698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:38656037 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGAGAATGAGAATTAGAACTTTT[A/G]TGAAGTGTTTATTCCCTAGCTATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPNE8 PubMed Links

Gene Details

Gene
CPNE8
Gene Name
copine 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153634.2 Intron NP_705898.1
XM_011537951.2 Intron XP_011536253.1
XM_011537952.2 Intron XP_011536254.1
XM_017018851.1 Intron XP_016874340.1
XM_017018852.1 Intron XP_016874341.1

View Full Product Details