Product Details

SNP ID
rs11551823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56418243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATCAGAGCCTTGCTCTCCAGG[G/T]ACTTTAGGCTGCAGCTCTGGCTGCA
Phenotype
MIM: 603887
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TIMELESS PubMed Links

Gene Details

Gene
TIMELESS
Gene Name
timeless circadian clock
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011538939.2 3488 Silent Mutation XP_011537241.1

View Full Product Details