Product Details

SNP ID
rs7615073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196877691 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAACTGTCTTGAACTAGTAGTTT[A/G]TAAGGTGTTTGACAAGTATAGAAGA
Phenotype
MIM: 612845
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SENP5 PubMed Links

Gene Details

Gene
SENP5
Gene Name
SUMO1/sentrin specific peptidase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308045.1 Intron NP_001294974.1
NM_152699.4 Intron NP_689912.2
XM_005269310.2 Intron XP_005269367.1
XM_005269312.2 Intron XP_005269369.1
XM_011512544.2 Intron XP_011510846.1
XM_011512545.2 Intron XP_011510847.1
XM_017005870.1 Intron XP_016861359.1

View Full Product Details