Product Details

SNP ID
rs7908923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95671100 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGTTATAGATAGGCAAAATTAGA[G/T]AATTCACTGTTTATTTAGTGTGTGC
Phenotype
MIM: 613847
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TCTN3 PubMed Links

Gene Details

Gene
TCTN3
Gene Name
tectonic family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143973.1 Intron NP_001137445.1
NM_015631.5 Intron NP_056446.4
XM_005269690.1 Intron XP_005269747.1
XM_011539627.1 Intron XP_011537929.1
XM_011539628.1 Intron XP_011537930.1

View Full Product Details