Product Details
- SNP ID
-
rs1378059
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:66595381 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACTTCCTGTACTACTTGCTTGCGA[A/G]GAGAGCAGACAGCTGGCCTTTCTCT
- Phenotype
-
MIM: 603864
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC87
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74720222] are located under a probe and SNP(s) [rs76681563] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC87
- Gene Name
- coiled-coil domain containing 87
There are no transcripts associated with this gene.
- Gene
- CCS
- Gene Name
- copper chaperone for superoxide dismutase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005125.1 |
|
Intron |
|
|
NP_005116.1 |
View Full Product Details