Product Details
- SNP ID
-
rs7842440
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:144280555 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTCCGGCACAGGCCAAACTCTCC[G/T]TAAATTATCTGGGAAGGCCGGGCCC
- Phenotype
-
MIM: 610596
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BOP1
PubMed Links
Gene Details
- Gene
- BOP1
- Gene Name
- block of proliferation 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015201.4 |
|
Intron |
|
|
NP_056016.1 |
View Full Product Details