Product Details

SNP ID
rs8052503
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1352142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAGCGAAGATGAAGGTGGTGGA[C/G]GAGCCCAACGCGTTTGGGTGAGCAG
Phenotype
MIM: 604009 MIM: 607838
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
BAIAP3 PubMed Links

Gene Details

Gene
BAIAP3
Gene Name
BAI1 associated protein 3
There are no transcripts associated with this gene.

Gene
GNPTG
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032520.4 128 Missense Mutation GAC,GAG D31E NP_115909.1
XM_017023782.1 128 Missense Mutation GAC,GAG D31E XP_016879271.1
XM_017023783.1 128 Intron XP_016879272.1
Gene
TSR3
Gene Name
TSR3, acp transferase ribosome maturation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001410.2 128 Intron NP_001001410.1

View Full Product Details