Product Details

SNP ID
rs11554987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1352243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTTGCCGCTTCCCGTAGGGTG[A/G]ACAACCCGTTCTTGCCTCAGGCCAG
Phenotype
MIM: 604009 MIM: 607838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAIAP3 PubMed Links

Gene Details

Gene
BAIAP3
Gene Name
BAI1 associated protein 3
There are no transcripts associated with this gene.

Gene
GNPTG
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032520.4 150 Missense Mutation AAC,GAC N39D NP_115909.1
XM_017023782.1 150 Missense Mutation AAC,GAC N39D XP_016879271.1
XM_017023783.1 150 Intron XP_016879272.1
Gene
TSR3
Gene Name
TSR3, acp transferase ribosome maturation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001410.2 150 Intron NP_001001410.1

View Full Product Details