Product Details

SNP ID
rs11557718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:636123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCAGCGCTGGTCCACGTCCGA[C/G]GGCTGCCACTCGGCCAGGGGGAAGG
Phenotype
MIM: 608021
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100287175 PubMed Links

Gene Details

Gene
LOC100287175
Gene Name
uncharacterized LOC100287175
There are no transcripts associated with this gene.

Gene
MCRIP2
Gene Name
MAPK regulated corepressor interacting protein 2
There are no transcripts associated with this gene.

Gene
METTL26
Gene Name
methyltransferase like 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040160.2 242 Silent Mutation CCC,CCG P56P NP_001035250.1
NM_001040161.2 242 Silent Mutation CCC,CCG P56P NP_001035251.1
NM_001040162.2 242 Silent Mutation CCC,CCG P56P NP_001035252.1
NM_001040165.2 242 Silent Mutation CCC,CCG P56P NP_001035255.1
NM_001288710.1 242 Silent Mutation CCC,CCG P56P NP_001275639.1
NM_032366.4 242 Silent Mutation CCC,CCG P56P NP_115742.3
XM_011522713.2 242 Silent Mutation CCC,CCG P56P XP_011521015.1
XM_011522714.2 242 Silent Mutation CCC,CCG P56P XP_011521016.1
Gene
RAB40C
Gene Name
RAB40C, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
WFIKKN1
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1
There are no transcripts associated with this gene.

View Full Product Details