Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286408.1 | 7691 | Intron | NP_001273337.1 | ||
NM_001286409.1 | 7691 | Intron | NP_001273338.1 | ||
NM_198081.4 | 7691 | Intron | NP_932347.2 | ||
XM_011535703.2 | 7691 | UTR 3 | XP_011534005.1 | ||
XM_011535704.2 | 7691 | UTR 3 | XP_011534006.1 | ||
XM_011535706.1 | 7691 | UTR 3 | XP_011534008.1 | ||
XM_011535707.2 | 7691 | Intron | XP_011534009.1 | ||
XM_011535709.2 | 7691 | UTR 3 | XP_011534011.1 | ||
XM_011535710.2 | 7691 | UTR 3 | XP_011534012.1 | ||
XM_017010677.1 | 7691 | UTR 3 | XP_016866166.1 | ||
XM_017010678.1 | 7691 | UTR 3 | XP_016866167.1 | ||
XM_017010679.1 | 7691 | UTR 3 | XP_016866168.1 | ||
XM_017010680.1 | 7691 | UTR 3 | XP_016866169.1 | ||
XM_017010681.1 | 7691 | UTR 3 | XP_016866170.1 | ||
XM_017010682.1 | 7691 | UTR 3 | XP_016866171.1 | ||
XM_017010683.1 | 7691 | UTR 3 | XP_016866172.1 | ||
XM_017010684.1 | 7691 | Intron | XP_016866173.1 | ||
XM_017010685.1 | 7691 | UTR 3 | XP_016866174.1 | ||
XM_017010686.1 | 7691 | UTR 3 | XP_016866175.1 | ||
XM_017010687.1 | 7691 | UTR 3 | XP_016866176.1 | ||
XM_017010688.1 | 7691 | Intron | XP_016866177.1 |