Product Details

SNP ID
rs1133893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8422000 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGCTCCTCTCCACCGTCATCCGT[A/G]CCTTGGACACACCGAGGTGAGTGGT
Phenotype
MIM: 613332
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARCH2 PubMed Links

Gene Details

Gene
MARCH2
Gene Name
membrane associated ring-CH-type finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005415.1 330 Missense Mutation ACC,GCC T54A NP_001005415.1
NM_001005416.1 330 Missense Mutation ACC,GCC T54A NP_001005416.1
NM_016496.4 330 Missense Mutation ACC,GCC T54A NP_057580.3
XM_006722763.3 330 Missense Mutation ACC,GCC T54A XP_006722826.1
XM_011528046.2 330 Missense Mutation ACC,GCC T54A XP_011526348.1
XM_011528047.2 330 Missense Mutation ACC,GCC T54A XP_011526349.1
XM_017026853.1 330 Missense Mutation ACC,GCC T54A XP_016882342.1
XM_017026854.1 330 Silent Mutation GTA,GTG V10V XP_016882343.1

View Full Product Details