Product Details
- SNP ID
-
rs12032405
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:1452045 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCTTTTTCTGCGGCTTCTTCTCA[A/G]CAACTTGTCAATGAGGATTTACGGA
- Phenotype
-
MIM: 611901
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATAD3C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112072883] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATAD3C
- Gene Name
- ATPase family, AAA domain containing 3C
- Gene
- VWA1
- Gene Name
- von Willebrand factor A domain containing 1
There are no transcripts associated with this gene.
View Full Product Details