Product Details

SNP ID
rs35836460
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:196651923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGCAGTGAGAATTGGGTTTAAC[C/T]TCTGGCATTTCTGGGCTTGTGGCTT
Phenotype
MIM: 134370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFH PubMed Links

Gene Details

Gene
CFH
Gene Name
complement factor H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000186.3 46 UTR 5 NP_000177.2
NM_001014975.2 46 UTR 5 NP_001014975.1
XM_017001108.1 46 Intron XP_016856597.1

View Full Product Details