Product Details

SNP ID
rs1064651
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:155235727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATGGGCTGTTTGTAAAACGTGT[C/G]CTTGGTGATGTCTACAATGATGGGA
Phenotype
MIM: 606463
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GBA PubMed Links

Gene Details

Gene
GBA
Gene Name
glucosylceramidase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000157.3 1767 Missense Mutation CAC,GAC H448D NP_000148.2
NM_001005741.2 1767 Missense Mutation CAC,GAC H448D NP_001005741.1
NM_001005742.2 1767 Missense Mutation CAC,GAC H448D NP_001005742.1
NM_001171811.1 1767 Missense Mutation CAC,GAC H361D NP_001165282.1
NM_001171812.1 1767 Missense Mutation CAC,GAC H399D NP_001165283.1
Gene
GBAP1
Gene Name
glucosylceramidase beta pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details