Product Details

SNP ID
rs28463524
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:156198242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTTCTAGTATCTTACAGAACA[C/G]CGGTGGGATACACTGGAGCACATCA
Phenotype
MIM: 610824
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A44 PubMed Links

Gene Details

Gene
SLC25A44
Gene Name
solute carrier family 25 member 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286184.1 Intron NP_001273113.1
NM_014655.3 Intron NP_055470.1
XM_006711657.3 Intron XP_006711720.1
XM_011510180.1 Intron XP_011508482.1
XM_011510181.1 Intron XP_011508483.1
XM_017002904.1 Intron XP_016858393.1
XM_017002905.1 Intron XP_016858394.1
XM_017002906.1 Intron XP_016858395.1
XM_017002907.1 Intron XP_016858396.1
XM_017002908.1 Intron XP_016858397.1

View Full Product Details