Product Details

SNP ID
rs28624719
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:156309754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCCTGAATGTAAAATGCAGTT[C/T]TCAGCCAGGTGTGGTGGCTCACACC
Phenotype
MIM: 600114
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCT3 PubMed Links

Gene Details

Gene
CCT3
Gene Name
chaperonin containing TCP1 subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008800.2 Intron NP_001008800.1
NM_005998.4 Intron NP_005989.3

View Full Product Details