Product Details

SNP ID
rs34861398
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:159172360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGTATGCAAACATTTCTGCTC[C/T]CCCGCCCGTCCCCCACCACCCGCGG
Phenotype
MIM: 609743
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CADM3 PubMed Links

Gene Details

Gene
CADM3
Gene Name
cell adhesion molecule 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127173.1 Intron NP_001120645.1
NM_021189.3 Intron NP_067012.1

View Full Product Details