Product Details

SNP ID
rs34439705
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:159176090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCCTACTCCAGTGGAACTGCAT[A/G]CTGGCTCAACAATCCCTTTTTTGTC
Phenotype
MIM: 609743
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CADM3 PubMed Links

Gene Details

Gene
CADM3
Gene Name
cell adhesion molecule 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127173.1 Intron NP_001120645.1
NM_021189.3 Intron NP_067012.1

View Full Product Details