Product Details

SNP ID
rs28904906
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:27892636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTTTCCAGCCACACACTGAAAC[C/T]CCGTTGGTCCCTCTCAGGGAAGTAA
Phenotype
MIM: 179836
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPA2 PubMed Links

Gene Details

Gene
RPA2
Gene Name
replication protein A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286076.1 Intron NP_001273005.1
NM_001297558.1 Intron NP_001284487.1
NM_002946.4 Intron NP_002937.1
Gene
THEMIS2
Gene Name
thymocyte selection associated family member 2
There are no transcripts associated with this gene.

View Full Product Details