Product Details

SNP ID
rs28970015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233677390 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAACCCGTGTGTAGTAGAAAATCC[G/T]CATAGAACTTTTGATTCTCCAAAAA
Phenotype
MIM: 606435 MIM: 606432 MIM: 606433 MIM: 606434
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UGT1A10 PubMed Links

Gene Details

Gene
UGT1A10
Gene Name
UDP glucuronosyltransferase family 1 member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019075.2 Intron NP_061948.1
Gene
UGT1A7
Gene Name
UDP glucuronosyltransferase family 1 member A7
There are no transcripts associated with this gene.

Gene
UGT1A8
Gene Name
UDP glucuronosyltransferase family 1 member A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019076.4 Intron NP_061949.3
Gene
UGT1A9
Gene Name
UDP glucuronosyltransferase family 1 member A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021027.2 Intron NP_066307.1

View Full Product Details