Product Details

SNP ID
rs28510539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202390018 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTATACAAAAAAAAAAAAAAATC[A/G]TCTCGTGTTTCCTTAATCCCATTTC
Phenotype
MIM: 600799
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMPR2 PubMed Links

Gene Details

Gene
BMPR2
Gene Name
bone morphogenetic protein receptor type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204.6 Intron NP_001195.2
XM_011511687.1 Intron XP_011509989.1

View Full Product Details