Product Details
- SNP ID
-
rs34964084
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:8592850 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCCCACCATGCCGCCCCCGCTGC[C/T]GCTGCTGCTCCTTACAGTCCTGGTC
- Phenotype
-
MIM: 603105
MIM: 606921
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CPZ
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2302583,rs76756393] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPZ
- Gene Name
- carboxypeptidase Z
- Gene
- GPR78
- Gene Name
- G protein-coupled receptor 78
There are no transcripts associated with this gene.
View Full Product Details