Product Details

SNP ID
rs17885864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:41745882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCGAAGATAGGACGCTGGCGAA[G/T]GGACCCCCAAGCGAATCCGGGATGG
Phenotype
MIM: 603851
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PHOX2B PubMed Links

Gene Details

Gene
PHOX2B
Gene Name
paired like homeobox 2b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003924.3 1230 Silent Mutation NP_003915.2

View Full Product Details