Product Details

SNP ID
rs28453134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:52989557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAGCGGGGATTTCGCTCTGCAT[A/C]GGCTGCAGGAGGGACCCGCGCGGCT
Phenotype
MIM: 192974
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ITGA2 PubMed Links

Gene Details

Gene
ITGA2
Gene Name
integrin subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002203.3 Intron NP_002194.2

View Full Product Details