Product Details

SNP ID
hCV58436561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:53098067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAAAAAAGAAAATACTCTTTTT[A/T]AACTTTTGAAATCATGGGTTTTAGT
Phenotype
MIM: 192974 MIM: 603708
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ITGA2 PubMed Links

Gene Details

Gene
ITGA2
Gene Name
integrin subunit alpha 2
There are no transcripts associated with this gene.

Gene
MOCS2
Gene Name
molybdenum cofactor synthesis 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004531.4 1332 UTR 3 NP_004522.1
NM_176806.3 1332 UTR 3 NP_789776.1

View Full Product Details