Product Details

SNP ID
rs28604505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:90409240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGTTTTTGTTATTTCCAAAAT[G/T]TATAGGTGGGTGATTGCATTAGCAG
Phenotype
MIM: 602907
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CETN3 PubMed Links

Gene Details

Gene
CETN3
Gene Name
centrin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297765.1 Intron NP_001284694.1
NM_001297768.1 Intron NP_001284697.1
NM_004365.3 Intron NP_004356.2
Gene
LOC731157
Gene Name
uncharacterized LOC731157
There are no transcripts associated with this gene.

View Full Product Details