Product Details

SNP ID
rs707784
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32553914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAAGGAAGAGCAGGCCCAGCAC[A/G]AAGCCCCCGACTCCACTCAGCATCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HLA-DRB6 PubMed Links
Additional Information
For this assay, SNP(s) [rs538116343] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HLA-DRB6
Gene Name
major histocompatibility complex, class II, DR beta 6 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details