Product Details

SNP ID
rs28475089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16795496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGGAATGCAAGTCCTTAACAAC[A/T]GCAAAGTTCAAATAAGCATATAGTA
Phenotype
MIM: 606358
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AGR2 PubMed Links

Gene Details

Gene
AGR2
Gene Name
anterior gradient 2, protein disulphide isomerase family member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006408.3 Intron NP_006399.1
XM_005249581.4 Intron XP_005249638.1

View Full Product Details