Product Details
- SNP ID
-
rs11548772
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:25123969 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTTACCTTTGTTCTTATTGGCGG[C/G]TGTGTAAGAGTATCCAGGGGCCTGA
- Phenotype
-
MIM: 123970
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CYCS
PubMed Links
Gene Details
- Gene
- CYCS
- Gene Name
- cytochrome c, somatic
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018947.5 |
321 |
Missense Mutation |
CCC,GCC |
P51A |
NP_061820.1 |
View Full Product Details