Product Details

SNP ID
rs28646224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:4900799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCTCCTGCCGCCTCCTCTCCAC[A/G]CCTTGCTCTTCCTGCTAGGGTTGAC
Phenotype
MIM: 614581
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMD2 PubMed Links

Gene Details

Gene
MMD2
Gene Name
monocyte to macrophage differentiation associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100600.1 Intron NP_001094070.1
NM_001270375.1 Intron NP_001257304.1
NM_198403.3 Intron NP_940685.3

View Full Product Details