Product Details

SNP ID
hCV58966677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140456191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACATTTCACACAGACTCCTTTTT[C/T]TTTCGTTTAAATAAATTTATTTCTT
Phenotype
MIM: 607754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MKRN1 PubMed Links

Gene Details

Gene
MKRN1
Gene Name
makorin ring finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145125.1 1672 UTR 3 NP_001138597.1
NM_001291663.1 1672 UTR 3 NP_001278592.1
NM_013446.3 1672 Intron NP_038474.2

View Full Product Details