Product Details
- SNP ID
-
hCV58972431
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149003061 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATCAGATGTTTTCTAACGCAGTT[C/T]TTAACGTTTTTAATCGTGTTTTATT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PDIA4
PubMed Links
Gene Details
- Gene
- PDIA4
- Gene Name
- protein disulfide isomerase family A member 4
There are no transcripts associated with this gene.
View Full Product Details