Product Details

SNP ID
rs28740842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:97738034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTACTTAAAATAATAACATAATT[A/T]TAAGTAGTGATAAAAATCACTGTAG
Phenotype
MIM: 162320
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TAC1 PubMed Links

Gene Details

Gene
TAC1
Gene Name
tachykinin precursor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003182.2 Intron NP_003173.1
NM_013996.2 Intron NP_054702.1
NM_013997.2 Intron NP_054703.1
NM_013998.2 Intron NP_054704.1

View Full Product Details