Product Details
- SNP ID
-
rs28954088
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:128245261 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTTCCCCTCTTTCCCCTGGGGAA[C/T]GCCTGTCCTTCAGGCCTCAGTTCAC
- Phenotype
-
MIM: 164160
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LEP
PubMed Links
Gene Details
- Gene
- LEP
- Gene Name
- leptin
View Full Product Details