Product Details

SNP ID
rs11545283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:134527637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGCACTTTGGAAGGCCGAGGT[A/G]GGCGGATCACCTGAGCTCAGGAGTT
Phenotype
MIM: 604707
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKR1B10 PubMed Links

Gene Details

Gene
AKR1B10
Gene Name
aldo-keto reductase family 1 member B10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020299.4 46 UTR 5 NP_064695.3
XM_011516416.1 46 UTR 5 XP_011514718.1
XM_011516417.2 46 UTR 5 XP_011514719.1

View Full Product Details