Product Details

SNP ID
rs28589845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37935110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGCCGTGGGTCCCACTCTGCTC[A/G]GTGATGTGACCCCAGGACCCAGGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOT1L1 PubMed Links

Gene Details

Gene
GOT1L1
Gene Name
glutamic-oxaloacetic transaminase 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152413.2 1415 Silent Mutation ACC,ACT T345T NP_689626.2
XM_005273399.3 1415 Silent Mutation ACC,ACT T344T XP_005273456.1
XM_006716285.3 1415 Silent Mutation ACC,ACT T314T XP_006716348.1
XM_011544391.2 1415 Silent Mutation ACC,ACT T163T XP_011542693.1

View Full Product Details