Product Details

SNP ID
hCV59228942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:130052386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTATTTGCAGAACATAACTTTCC[C/T]CTCTGGGGGAAAAAGAACTAATTTG
Phenotype
MIM: 605953
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASAP1 PubMed Links

Gene Details

Gene
ASAP1
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001247996.1 6065 UTR 3 NP_001234925.1
NM_018482.3 6065 UTR 3 NP_060952.2

View Full Product Details