Product Details

SNP ID
rs34258920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:63066063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGATTAGACTTACTCGAAATAC[A/G]TCATAAGCTGTAAAAACTTTGGGGT
Phenotype
MIM: 600415
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTPA PubMed Links

Gene Details

Gene
TTPA
Gene Name
alpha tocopherol transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000370.3 425 Silent Mutation NP_000361.1
XM_006716468.3 425 Intron XP_006716531.1

View Full Product Details