Product Details

SNP ID
rs28681637
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:49911309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAAGAAGCTCTGGATTTTCCA[A/C]GGTACAAATCTATATATTTTTTCTC
Phenotype
MIM: 608714
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
SNTG1 PubMed Links

Gene Details

Gene
SNTG1
Gene Name
syntrophin gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287813.2 Intron NP_001274742.1
NM_001287814.2 Intron NP_001274743.1
NM_001321773.1 Intron NP_001308702.1
NM_001321775.1 Intron NP_001308704.1
NM_001321776.1 Intron NP_001308705.1
NM_001321777.1 Intron NP_001308706.1
NM_001321778.1 Intron NP_001308707.1
NM_018967.4 Intron NP_061840.1
XM_011517548.2 Intron XP_011515850.1
XM_017013579.1 Intron XP_016869068.1
XM_017013580.1 Intron XP_016869069.1
XM_017013581.1 Intron XP_016869070.1
XM_017013582.1 Intron XP_016869071.1

View Full Product Details