Product Details

SNP ID
rs28475207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:79922459 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTAGACCTGGAAATTGCTAAGTGA[A/G]TATATTTTGTGTTCTCATCATCAAA
Phenotype
MIM: 611990
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS28 PubMed Links

Gene Details

Gene
MRPS28
Gene Name
mitochondrial ribosomal protein S28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014018.2 Intron NP_054737.1

View Full Product Details