Product Details

SNP ID
rs28410532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:46002136 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATTGATGCGTCAGTGCATGAAGT[T/G]GGGGGTGGTCACCAGCATCTAACTC
Phenotype
MIM: 601984 MIM: 605034
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
NCOA4 PubMed Links

Gene Details

Gene
NCOA4
Gene Name
nuclear receptor coactivator 4
There are no transcripts associated with this gene.

Gene
TIMM23
Gene Name
translocase of inner mitochondrial membrane 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006327.3 Intron NP_006318.1
XM_011539098.1 Intron XP_011537400.1
XM_011539099.1 Intron XP_011537401.1

View Full Product Details