Product Details
- SNP ID
-
hCV59735550
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:22827827 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAACTTGATCTTTAATCTTACCTC[T/C]TTTTGTCTTCTCTCTGCAGCCTCTG
- Phenotype
-
MIM: 602835
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAS2
PubMed Links
Gene Details
- Gene
- GAS2
- Gene Name
- growth arrest specific 2
There are no transcripts associated with this gene.
- Gene
- SVIP
- Gene Name
- small VCP interacting protein
There are no transcripts associated with this gene.
View Full Product Details