Product Details

SNP ID
rs28362803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14837190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACACAAGCTTCTAATTGATAA[A/G]TTTCTGGGAAAAATTTAGTTACGGT
Phenotype
MIM: 110600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ART4 PubMed Links

Gene Details

Gene
ART4
Gene Name
ADP-ribosyltransferase 4 (Dombrock blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021071.2 Intron NP_066549.2

View Full Product Details