Product Details

SNP ID
rs28820735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123393960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGGGGACATATGTTTTTAATT[C/T]TCTGGAATAGATGCCTAGGAGTGAA
Phenotype
MIM: 607240
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KMT5A PubMed Links

Gene Details

Gene
KMT5A
Gene Name
lysine methyltransferase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324504.1 Intron NP_001311433.1
NM_001324505.1 Intron NP_001311434.1
NM_001324506.1 Intron NP_001311435.1
NM_020382.4 Intron NP_065115.3
XM_006719394.3 Intron XP_006719457.1
XM_011538333.2 Intron XP_011536635.1

View Full Product Details