Product Details

SNP ID
hCV59935966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:130873095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGACACAGCCGGCCAGGAGAAA[C/T]TCGGTGGACTGAGAGATGGCTATTA
Phenotype
MIM: 601179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAN PubMed Links

Gene Details

Gene
RAN
Gene Name
RAN, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300796.1 303 UTR 5 NP_001287725.1
NM_001300797.1 303 UTR 5 NP_001287726.1
NM_006325.4 303 Missense Mutation CTC,TTC L72F NP_006316.1

View Full Product Details