Product Details

SNP ID
rs11542827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40038900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTCTTCAAGGTGATATAGACGC[C/T]GCCCGACGTCCGGCACTTCTGGAAA
Phenotype
MIM: 609280 MIM: 600708
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2AK4 PubMed Links

Gene Details

Gene
EIF2AK4
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 4
There are no transcripts associated with this gene.

Gene
SRP14
Gene Name
signal recognition particle 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309434.1 159 UTR 5 NP_001296363.1
NM_003134.5 159 Missense Mutation AGC,GGC S25G NP_003125.3
Gene
SRP14-AS1
Gene Name
SRP14 antisense RNA1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details